Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116949 MIM: 117140 MIM: 610674 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C20orf27 - chromosome 20 open reading frame 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDC25B - cell division cycle 25B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CENPB - centromere protein B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPEF1 - sperm flagellar 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015417.4 | 1271 | UTR 3 | NP_056232.2 | |||
XM_005260683.4 | 1271 | UTR 3 | XP_005260740.1 |