Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF639 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF639 - zinc finger protein 639 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303425.1 | 74 | Intron | NP_001290354.1 | |||
NM_001303426.1 | 74 | Intron | NP_001290355.1 | |||
NM_016331.2 | 74 | Intron | NP_057415.1 | |||
XM_017006550.1 | 74 | UTR 5 | XP_016862039.1 | |||
XM_017006551.1 | 74 | Intron | XP_016862040.1 | |||
XM_017006552.1 | 74 | Intron | XP_016862041.1 | |||
XM_017006553.1 | 74 | UTR 5 | XP_016862042.1 |
Set Membership: |
HapMap |