Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTATCCCTCTGTGTTTTCTTTG[G/T]ATAAAAATATTAAAATTTGGGTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613921 MIM: 609360 | ||||||||||||||||||||
Literature Links: |
LIPJ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIPJ - lipase family member J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010939.2 | Intron | NP_001010939.2 | ||||
XM_005269542.3 | Intron | XP_005269599.1 | ||||
XM_006717635.2 | Intron | XP_006717698.1 | ||||
XM_011539314.2 | Intron | XP_011537616.1 | ||||
XM_011539315.2 | Intron | XP_011537617.1 | ||||
XM_011539317.2 | Intron | XP_011537619.1 | ||||
XM_011539318.2 | Intron | XP_011537620.1 | ||||
XM_011539319.1 | Intron | XP_011537621.1 | ||||
XM_011539320.2 | Intron | XP_011537622.1 | ||||
XM_017015740.1 | Intron | XP_016871229.1 |
RNLS - renalase, FAD dependent amine oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |