Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTCAAAGGGCAAGGATCATGT[A/T]GTGTTTACCTTTGTATCCTCAACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609953 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAVER2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAVER2 - ribonucleoprotein, PTB binding 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018211.3 | Intron | NP_060681.2 | ||||
XM_006710738.3 | Intron | XP_006710801.2 | ||||
XM_011541706.2 | Intron | XP_011540008.1 |
Set Membership: |
HapMap |