Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACTTATCTTTTTTCTTTTGGCC[A/T]TGAGTGGAAGGTAAGCTGAGGTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604586 | ||||||||||||||||||||
Literature Links: |
STXBP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STXBP5 - syntaxin binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127715.2 | Intron | NP_001121187.1 | ||||
NM_139244.4 | Intron | NP_640337.3 | ||||
XM_006715339.2 | Intron | XP_006715402.1 | ||||
XM_011535453.1 | Intron | XP_011533755.1 | ||||
XM_017010267.1 | Intron | XP_016865756.1 | ||||
XM_017010268.1 | Intron | XP_016865757.1 |
STXBP5-AS1 - STXBP5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |