Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608009 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK5 - adenylate kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012093.3 | Intron | NP_036225.2 | ||||
NM_174858.2 | Intron | NP_777283.1 | ||||
XM_005270739.4 | Intron | XP_005270796.1 | ||||
XM_006710572.3 | Intron | XP_006710635.1 | ||||
XM_017001008.1 | Intron | XP_016856497.1 | ||||
XM_017001009.1 | Intron | XP_016856498.1 | ||||
XM_017001010.1 | Intron | XP_016856499.1 | ||||
XM_017001011.1 | Intron | XP_016856500.1 | ||||
XM_017001012.1 | Intron | XP_016856501.1 |
Set Membership: |
HapMap |