Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAT1 - branched chain amino acid transaminase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178091.1 | 8953 | UTR 3 | NP_001171562.1 | |||
NM_001178092.1 | 8953 | UTR 3 | NP_001171563.1 | |||
NM_001178093.1 | 8953 | UTR 3 | NP_001171564.1 | |||
NM_001178094.1 | 8953 | UTR 3 | NP_001171565.1 | |||
NM_005504.6 | 8953 | UTR 3 | NP_005495.2 | |||
XM_017019768.1 | 8953 | Intron | XP_016875257.1 |