Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTAATCAGGGAACTAATAAAT[G/T]CTTAGTTCATGGGAAAACTTCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605454 MIM: 607613 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCB10 - ATP binding cassette subfamily B member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012089.2 | 4259 | Intron | NP_036221.2 | |||
XM_011544135.2 | 4259 | UTR 3 | XP_011542437.1 | |||
XM_011544136.2 | 4259 | Intron | XP_011542438.1 |
LOC101927478 - uncharacterized LOC101927478 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP133 - nucleoporin 133 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |