Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGAAACTCCGTCTAGAAGCCT[A/G]CAGGAGAGGTGGGAAGGCTGGACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf61 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf61 - chromosome 1 open reading frame 61 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEF2D - myocyte enhancer factor 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271629.1 | 4557 | UTR 3 | NP_001258558.1 | |||
NM_005920.3 | 4557 | UTR 3 | NP_005911.1 | |||
XM_005245169.4 | 4557 | UTR 3 | XP_005245226.1 | |||
XM_005245170.3 | 4557 | UTR 3 | XP_005245227.1 | |||
XM_006711330.3 | 4557 | UTR 3 | XP_006711393.1 | |||
XM_006711332.3 | 4557 | UTR 3 | XP_006711395.1 | |||
XM_006711333.2 | 4557 | UTR 3 | XP_006711396.1 | |||
XM_006711334.3 | 4557 | UTR 3 | XP_006711397.1 | |||
XM_011509569.2 | 4557 | UTR 3 | XP_011507871.1 | |||
XM_017001314.1 | 4557 | UTR 3 | XP_016856803.1 | |||
XM_017001315.1 | 4557 | UTR 3 | XP_016856804.1 |