Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAACTTTCTGAATTAACGTTAT[G/T]TAAAAGGAAATGTAGATGTTATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFHR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFHR5 - complement factor H related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030787.3 | 2640 | UTR 3 | NP_110414.1 | |||
XM_011510020.2 | 2640 | UTR 3 | XP_011508322.1 |