Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATAAAATTCAAAGAAACAAAT[A/G]ATATGTGTTGGCATTTCTTGATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609984 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCTL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LCTL - lactase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278562.1 | 3290 | Intron | NP_001265491.1 | |||
NM_207338.3 | 3290 | Intron | NP_997221.2 | |||
XM_017021999.1 | 3290 | Intron | XP_016877488.1 |
ZWILCH - zwilch kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287821.1 | 3290 | UTR 3 | NP_001274750.1 | |||
NM_001287822.1 | 3290 | UTR 3 | NP_001274751.1 | |||
NM_001287823.1 | 3290 | UTR 3 | NP_001274752.1 | |||
NM_017975.4 | 3290 | UTR 3 | NP_060445.3 |
Set Membership: |
HapMap |