Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGAGAAGTCCTGAGAGGTGGC[T/C]TCAGCAGGGAGGAGCAAGGTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607901 MIM: 605063 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FERMT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FERMT3 - fermitin family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031471.5 | Intron | NP_113659.3 | ||||
NM_178443.2 | Intron | NP_848537.1 | ||||
XM_011545294.2 | Intron | XP_011543596.1 | ||||
XM_011545295.2 | Intron | XP_011543597.1 | ||||
XM_017018398.1 | Intron | XP_016873887.1 | ||||
XM_017018399.1 | Intron | XP_016873888.1 |
STIP1 - stress induced phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |