Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172480 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSPH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PSPH - phosphoserine phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004577.3 | Intron | NP_004568.2 | ||||
XM_005271773.1 | Intron | XP_005271830.1 | ||||
XM_005271774.1 | Intron | XP_005271831.1 | ||||
XM_005271775.1 | Intron | XP_005271832.1 | ||||
XM_005271776.1 | Intron | XP_005271833.1 | ||||
XM_006715760.1 | Intron | XP_006715823.1 | ||||
XM_006715761.1 | Intron | XP_006715824.1 | ||||
XM_011515461.1 | Intron | XP_011513763.1 | ||||
XM_017012466.1 | Intron | XP_016867955.1 | ||||
XM_017012467.1 | Intron | XP_016867956.1 | ||||
XM_017012468.1 | Intron | XP_016867957.1 | ||||
XM_017012469.1 | Intron | XP_016867958.1 |
Set Membership: |
HapMap |