Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTAGGTCAGGAGAGATGAAATC[A/G]ACAACAGTCCGTTTGCTGCTTATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606229 MIM: 607268 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGO2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AGO2 - argonaute 2, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164623.1 | 12648 | Intron | NP_001158095.1 | |||
NM_012154.3 | 12648 | Intron | NP_036286.2 | |||
XM_011516965.2 | 12648 | UTR 3 | XP_011515267.1 | |||
XM_011516966.2 | 12648 | UTR 3 | XP_011515268.1 | |||
XM_011516968.2 | 12648 | UTR 3 | XP_011515270.2 | |||
XM_017013317.1 | 12648 | UTR 3 | XP_016868806.1 |
CHRAC1 - chromatin accessibility complex 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |