Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGTTCTGAGCTCTGCTGAAATA[C/T]GGGGATATAAAGGTCTTGCCTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602425 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAP3K4 - mitogen-activated protein kinase kinase kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291958.1 | Intron | NP_001278887.1 | ||||
NM_001301072.1 | Intron | NP_001288001.1 | ||||
NM_005922.3 | Intron | NP_005913.2 | ||||
NM_006724.3 | Intron | NP_006715.2 | ||||
XM_005266989.2 | Intron | XP_005267046.1 | ||||
XM_017010869.1 | Intron | XP_016866358.1 |
Set Membership: |
Validated |