Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTGTCAGAATCTTTAAGTCACA[C/T]TAACTTGTACTTCCAAAGAATAGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300724 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNKSR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CNKSR2 - connector enhancer of kinase suppressor of Ras 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168647.1 | Intron | NP_001162118.1 | ||||
NM_001168648.1 | Intron | NP_001162119.1 | ||||
NM_001168649.1 | Intron | NP_001162120.1 | ||||
NM_014927.3 | Intron | NP_055742.2 | ||||
XM_011545471.2 | Intron | XP_011543773.1 | ||||
XM_011545472.2 | Intron | XP_011543774.1 | ||||
XM_017029358.1 | Intron | XP_016884847.1 |