Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTCCCTAACCCTGGGGATCAC[A/G]CAGAGCCCACATTCCAGGTGACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf180 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C14orf180 - chromosome 14 open reading frame 180 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008404.2 | Intron | NP_001008404.1 | ||||
NM_001286399.1 | Intron | NP_001273328.1 | ||||
NM_001286400.1 | Intron | NP_001273329.1 | ||||
XM_005267638.3 | Intron | XP_005267695.1 | ||||
XM_011536764.2 | Intron | XP_011535066.1 |
TMEM179 - transmembrane protein 179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |