Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAAGAGGTCCAGGTTCTACCCC[C/G]ACCCACATCCCCTGAATCAGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP10 - FK506 binding protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NT5C3B - 5'-nucleotidase, cytosolic IIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052935.4 | 1350 | UTR 3 | NP_443167.4 | |||
XM_006721669.3 | 1350 | Intron | XP_006721732.1 | |||
XM_006721670.3 | 1350 | Intron | XP_006721733.1 | |||
XM_011524276.2 | 1350 | Intron | XP_011522578.1 | |||
XM_011524277.2 | 1350 | Intron | XP_011522579.1 | |||
XM_017024127.1 | 1350 | Intron | XP_016879616.1 |