Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607063 MIM: 608778 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FKBP10 - FK506 binding protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLHL10 - kelch like family member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NT5C3B - 5'-nucleotidase, cytosolic IIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052935.4 | Intron | NP_443167.4 | ||||
XM_006721669.3 | Intron | XP_006721732.1 | ||||
XM_006721670.3 | Intron | XP_006721733.1 | ||||
XM_011524276.2 | Intron | XP_011522578.1 | ||||
XM_011524277.2 | Intron | XP_011522579.1 | ||||
XM_017024127.1 | Intron | XP_016879616.1 |
Set Membership: |
HapMap |