Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTCCCGGGATAGGAGCAAGCCT[T/C]GCACTCACAAAAGCCTCTGTCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf51 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C17orf51 - chromosome 17 open reading frame 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113434.4 | 4923 | UTR 3 | NP_001106905.1 | |||
XM_005256621.3 | 4923 | Intron | XP_005256678.1 | |||
XM_006721514.2 | 4923 | Intron | XP_006721577.1 | |||
XM_011523822.2 | 4923 | Intron | XP_011522124.1 |