Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601545 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAFAH1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PAFAH1B1 - platelet activating factor acetylhydrolase 1b regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000430.3 | Intron | NP_000421.1 | ||||
XM_011523901.1 | Intron | XP_011522203.1 | ||||
XM_011523902.2 | Intron | XP_011522204.1 | ||||
XM_011523903.2 | Intron | XP_011522205.1 | ||||
XM_017024699.1 | Intron | XP_016880188.1 | ||||
XM_017024700.1 | Intron | XP_016880189.1 | ||||
XM_017024701.1 | Intron | XP_016880190.1 | ||||
XM_017024702.1 | Intron | XP_016880191.1 | ||||
XM_017024703.1 | Intron | XP_016880192.1 |