Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601545 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAFAH1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAFAH1B1 - platelet activating factor acetylhydrolase 1b regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000430.3 | 717 | Intron | NP_000421.1 | |||
XM_011523901.1 | 717 | Intron | XP_011522203.1 | |||
XM_011523902.2 | 717 | UTR 5 | XP_011522204.1 | |||
XM_011523903.2 | 717 | Intron | XP_011522205.1 | |||
XM_017024699.1 | 717 | Intron | XP_016880188.1 | |||
XM_017024700.1 | 717 | UTR 5 | XP_016880189.1 | |||
XM_017024701.1 | 717 | Intron | XP_016880190.1 | |||
XM_017024702.1 | 717 | Intron | XP_016880191.1 | |||
XM_017024703.1 | 717 | Missense Mutation | GGG,TGG | G,W 213 | XP_016880192.1 |