Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCTTTCATACAACCGAGATCAA[A/C]GTAAACACTGCCCAGGAAATATACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603741 MIM: 607206 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALOX12B - arachidonate 12-lipoxygenase, 12R type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ALOXE3 - arachidonate lipoxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165960.1 | Intron | NP_001159432.1 | ||||
NM_021628.2 | Intron | NP_067641.2 | ||||
XM_017024921.1 | Intron | XP_016880410.1 | ||||
XM_017024922.1 | Intron | XP_016880411.1 | ||||
XM_017024923.1 | Intron | XP_016880412.1 | ||||
XM_017024924.1 | Intron | XP_016880413.1 | ||||
XM_017024925.1 | Intron | XP_016880414.1 |
MIR4314 - microRNA 4314 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |