Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACACTGCCGAATGTCATCTCTT[C/G]TCATCTTTATCTCTATTCTTTGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602546 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C15orf32 - chromosome 15 open reading frame 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301106.1 | 3636 | Intron | NP_001288035.1 | |||
NM_153040.2 | 3636 | Intron | NP_694585.1 | |||
XM_011521257.2 | 3636 | UTR 5 | XP_011519559.1 |
LOC104613533 - uncharacterized LOC104613533 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST8SIA2 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006011.3 | 3636 | Intron | NP_006002.1 | |||
XM_017022642.1 | 3636 | Intron | XP_016878131.1 | |||
XM_017022643.1 | 3636 | Intron | XP_016878132.1 |
Set Membership: |
HapMap |