Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTACAGAAGGTACGAAATAAAC[A/G]CCGATCTCCTTTATTCTGATCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602416 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3S2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP3S2 - adaptor related protein complex 3 sigma 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005829.4 | 2965 | UTR 3 | NP_005820.1 |
C15orf38-AP3S2 - C15orf38-AP3S2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199058.1 | 2965 | UTR 3 | NP_001185987.1 |
Set Membership: |
HapMap |