Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATAAATGTCAAATGGAAAAAG[A/G]ACAGTGTAAAGTGAGATATCATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606654 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RXFP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RXFP1 - relaxin/insulin like family peptide receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253727.1 | Intron | NP_001240656.1 | ||||
NM_001253728.1 | Intron | NP_001240657.1 | ||||
NM_001253729.1 | Intron | NP_001240658.1 | ||||
NM_001253730.1 | Intron | NP_001240659.1 | ||||
NM_001253732.1 | Intron | NP_001240661.1 | ||||
NM_001253733.1 | Intron | NP_001240662.1 | ||||
NM_021634.3 | Intron | NP_067647.2 | ||||
XM_011532174.1 | Intron | XP_011530476.1 | ||||
XM_011532175.1 | Intron | XP_011530477.1 | ||||
XM_011532176.1 | Intron | XP_011530478.1 | ||||
XM_011532179.2 | Intron | XP_011530481.1 | ||||
XM_017008517.1 | Intron | XP_016864006.1 | ||||
XM_017008518.1 | Intron | XP_016864007.1 | ||||
XM_017008519.1 | Intron | XP_016864008.1 | ||||
XM_017008520.1 | Intron | XP_016864009.1 | ||||
XM_017008521.1 | Intron | XP_016864010.1 | ||||
XM_017008522.1 | Intron | XP_016864011.1 | ||||
XM_017008523.1 | Intron | XP_016864012.1 | ||||
XM_017008524.1 | Intron | XP_016864013.1 | ||||
XM_017008525.1 | Intron | XP_016864014.1 | ||||
XM_017008526.1 | Intron | XP_016864015.1 |