Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATATACATTTACACCTAATTTT[C/G]AGTAGCACACTAAATGATTCACATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT11 - CCR4-NOT transcription complex subunit 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF149 - ring finger protein 149 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173647.3 | 2456 | Intron | NP_775918.2 | |||
XM_005263920.2 | 2456 | Intron | XP_005263977.1 | |||
XM_005263921.3 | 2456 | UTR 3 | XP_005263978.1 | |||
XM_011510990.2 | 2456 | Intron | XP_011509292.1 |
SNORD89 - small nucleolar RNA, C/D box 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |