Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601487 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPARG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPARG - peroxisome proliferator activated receptor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005037.5 | Intron | NP_005028.4 | ||||
NM_015869.4 | Intron | NP_056953.2 | ||||
NM_138711.3 | Intron | NP_619725.2 | ||||
NM_138712.3 | Intron | NP_619726.2 | ||||
XM_011533841.2 | Intron | XP_011532143.1 | ||||
XM_011533842.2 | Intron | XP_011532144.1 | ||||
XM_011533843.2 | Intron | XP_011532145.1 | ||||
XM_011533844.1 | Intron | XP_011532146.1 |
Set Membership: |
HapMap |