Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACGCAGCTGTGGAAGAAAGAAA[G/T]CAGTTGTACTGACTGGTTTCACTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYSMD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYSMD4 - LysM domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284417.1 | Intron | NP_001271346.1 | ||||
NM_001284418.1 | Intron | NP_001271347.1 | ||||
NM_001284419.1 | Intron | NP_001271348.1 | ||||
NM_001284420.1 | Intron | NP_001271349.1 | ||||
NM_001284421.1 | Intron | NP_001271350.1 | ||||
NM_001284422.1 | Intron | NP_001271351.1 | ||||
NM_152449.3 | Intron | NP_689662.2 | ||||
XM_006720397.3 | Intron | XP_006720460.2 | ||||
XM_011521241.1 | Intron | XP_011519543.1 | ||||
XM_011521242.2 | Intron | XP_011519544.1 | ||||
XM_011521243.2 | Intron | XP_011519545.1 | ||||
XM_011521244.1 | Intron | XP_011519546.1 | ||||
XM_011521245.2 | Intron | XP_011519547.1 | ||||
XM_017021926.1 | Intron | XP_016877415.1 | ||||
XM_017021927.1 | Intron | XP_016877416.1 | ||||
XM_017021928.1 | Intron | XP_016877417.1 | ||||
XM_017021929.1 | Intron | XP_016877418.1 | ||||
XM_017021930.1 | Intron | XP_016877419.1 |
MEF2A - myocyte enhancer factor 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |