Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605487 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPP19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARPP19 - cAMP regulated phosphoprotein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306191.1 | 2431 | UTR 3 | NP_001293120.1 | |||
NM_001306195.1 | 2431 | Intron | NP_001293124.1 | |||
NM_001306196.1 | 2431 | UTR 3 | NP_001293125.1 | |||
NM_006628.5 | 2431 | UTR 3 | NP_006619.1 | |||
XM_017021868.1 | 2431 | Intron | XP_016877357.1 | |||
XM_017021869.1 | 2431 | UTR 3 | XP_016877358.1 | |||
XM_017021870.1 | 2431 | Intron | XP_016877359.1 | |||
XM_017021871.1 | 2431 | UTR 3 | XP_016877360.1 |