Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGCGGGGACAGCAGGGCTCCGG[G/C]CAAGGAGGTCTGGATGCTGAGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4E1B - eukaryotic translation initiation factor 4E family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099408.1 | 41 | Intron | NP_001092878.1 | |||
XM_011534499.2 | 41 | Intron | XP_011532801.1 |
TSPAN17 - tetraspanin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006616.2 | 41 | UTR 5 | NP_001006617.2 | |||
NM_012171.2 | 41 | UTR 5 | NP_036303.1 | |||
NM_130465.4 | 41 | UTR 5 | NP_569732.2 | |||
XM_005265876.3 | 41 | UTR 5 | XP_005265933.1 |