Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAAACGAGGCTGGTGAGGATTC[C/T]GTGATGATGGATGGAAAGCATGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602569 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EIF4E1B - eukaryotic translation initiation factor 4E family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4281 - microRNA 4281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNCB - synuclein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001502.2 | Intron | NP_001001502.1 | ||||
NM_001318034.1 | Intron | NP_001304963.1 | ||||
NM_001318035.1 | Intron | NP_001304964.1 | ||||
NM_001318036.1 | Intron | NP_001304965.1 | ||||
NM_001318037.1 | Intron | NP_001304966.1 | ||||
NM_003085.4 | Intron | NP_003076.1 | ||||
XM_006714914.3 | Intron | XP_006714977.1 | ||||
XM_006714915.3 | Intron | XP_006714978.1 | ||||
XM_006714916.2 | Intron | XP_006714979.1 | ||||
XM_011534640.2 | Intron | XP_011532942.1 |
Set Membership: |
Validated |