Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603127 MIM: 603659 MIM: 179618 | ||||||||||||||||||||
Literature Links: |
GAS7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAS7 - growth arrest specific 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLP2R - glucagon like peptide 2 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCVRN - recoverin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002903.2 | 655 | Missense Mutation | TGG,TGT | W,C 156 | NP_002894.1 |