Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611459 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC10A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC10A7 - solute carrier family 10 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029998.5 | 1651 | UTR 3 | NP_001025169.1 | |||
NM_001300842.2 | 1651 | UTR 3 | NP_001287771.1 | |||
NM_001317816.1 | 1651 | UTR 3 | NP_001304745.1 | |||
NM_001317817.1 | 1651 | Intron | NP_001304746.1 | |||
NM_001317818.1 | 1651 | Intron | NP_001304747.1 | |||
NM_032128.4 | 1651 | Intron | NP_115504.1 | |||
XM_011532311.2 | 1651 | UTR 3 | XP_011530613.1 | |||
XM_011532313.2 | 1651 | UTR 3 | XP_011530615.1 | |||
XM_011532314.2 | 1651 | UTR 3 | XP_011530616.1 | |||
XM_017008689.1 | 1651 | Intron | XP_016864178.1 | |||
XM_017008690.1 | 1651 | Intron | XP_016864179.1 | |||
XM_017008691.1 | 1651 | Intron | XP_016864180.1 | |||
XM_017008692.1 | 1651 | Intron | XP_016864181.1 |
Set Membership: |
HapMap Validated |