Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603475 MIM: 606023 MIM: 600044 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRD - chordin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2H - RNA polymerase II subunit H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THPO - thrombopoietin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000460.3 | 918 | Silent Mutation | ACA,ACT | T,T 213 | NP_000451.1 | |
NM_001177597.2 | 918 | Silent Mutation | ACA,ACT | T,T 209 | NP_001171068.1 | |
NM_001177598.2 | 918 | Missense Mutation | AAC,TAC | N,Y 208 | NP_001171069.1 | |
NM_001289997.1 | 918 | Missense Mutation | AAC,TAC | N,Y 175 | NP_001276926.1 | |
NM_001289998.1 | 918 | Silent Mutation | ACA,ACT | T,T 213 | NP_001276927.1 | |
NM_001290003.1 | 918 | Silent Mutation | ACA,ACT | T,T 353 | NP_001276932.1 | |
NM_001290022.1 | 918 | Silent Mutation | ACA,ACT | T,T 209 | NP_001276951.1 | |
NM_001290026.1 | 918 | Missense Mutation | AAC,TAC | N,Y 208 | NP_001276955.1 | |
NM_001290027.1 | 918 | Missense Mutation | AAC,TAC | N,Y 175 | NP_001276956.1 | |
NM_001290028.1 | 918 | Silent Mutation | ACA,ACT | T,T 213 | NP_001276957.1 | |
XM_017007107.1 | 918 | Missense Mutation | AAC,TAC | N,Y 311 | XP_016862596.1 |