Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606980 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COQ8A - coenzyme Q8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020247.4 | Intron | NP_064632.2 | ||||
XM_005273201.1 | Intron | XP_005273258.1 | ||||
XM_011544238.1 | Intron | XP_011542540.1 | ||||
XM_011544239.2 | Intron | XP_011542541.1 | ||||
XM_011544240.2 | Intron | XP_011542542.1 | ||||
XM_011544241.2 | Intron | XP_011542543.1 | ||||
XM_017001852.1 | Intron | XP_016857341.1 |