Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610837 MIM: 608846 MIM: 602950 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADM5 - adrenomedullin 5 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BCL2L12 - BCL2 like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CPT1C - carnitine palmitoyltransferase 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5088 - microRNA 5088 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT1 - protein arginine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207042.2 | Intron | NP_001193971.1 | ||||
NM_001536.5 | Intron | NP_001527.3 | ||||
NM_198318.4 | Intron | NP_938074.2 | ||||
XM_005258842.1 | Intron | XP_005258899.1 | ||||
XM_017026734.1 | Intron | XP_016882223.1 | ||||
XM_017026735.1 | Intron | XP_016882224.1 | ||||
XM_017026736.1 | Intron | XP_016882225.1 |
Set Membership: |
HapMap |