Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606436 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYT12 - synaptotagmin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177880.1 | Intron | NP_001171351.1 | ||||
NM_001318773.1 | Intron | NP_001305702.1 | ||||
NM_001318775.1 | Intron | NP_001305704.1 | ||||
NM_177963.3 | Intron | NP_808878.1 | ||||
XM_006718737.3 | Intron | XP_006718800.1 | ||||
XM_011545346.2 | Intron | XP_011543648.1 | ||||
XM_011545347.1 | Intron | XP_011543649.1 | ||||
XM_017018547.1 | Intron | XP_016874036.1 | ||||
XM_017018548.1 | Intron | XP_016874037.1 |
Set Membership: |
HapMap |