Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609651 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPHA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPHA2 - glycoprotein hormone alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAJIN - membrane anchored junction protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037225.2 | Intron | NP_001032302.1 | ||||
NM_001300803.1 | Intron | NP_001287732.1 | ||||
NM_001318808.1 | Intron | NP_001305737.1 | ||||
XM_005273918.2 | Intron | XP_005273975.1 | ||||
XM_006718502.2 | Intron | XP_006718565.1 | ||||
XM_006718503.3 | Intron | XP_006718566.1 | ||||
XM_006718504.2 | Intron | XP_006718567.1 | ||||
XM_006718505.2 | Intron | XP_006718568.1 | ||||
XM_011544940.2 | Intron | XP_011543242.1 | ||||
XM_011544941.2 | Intron | XP_011543243.1 |