Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165340 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SKIL PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SKIL - SKI-like proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145097.2 | Intron | NP_001138569.1 | ||||
NM_001145098.2 | Intron | NP_001138570.1 | ||||
NM_001248008.1 | Intron | NP_001234937.1 | ||||
NM_005414.4 | Intron | NP_005405.2 | ||||
XM_005247721.1 | Intron | XP_005247778.1 | ||||
XM_006713735.1 | Intron | XP_006713798.1 |