Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGCGAGCCCCCAGCGGTGTGCT[A/G]GGATGGAAAAGCAGGAGCCGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616485 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZBTB21 - zinc finger and BTB domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098402.1 | Intron | NP_001091872.1 | ||||
NM_001098403.1 | Intron | NP_001091873.1 | ||||
NM_001320729.1 | Intron | NP_001307658.1 | ||||
NM_001320731.1 | Intron | NP_001307660.1 | ||||
NM_020727.4 | Intron | NP_065778.3 | ||||
XM_005261121.3 | Intron | XP_005261178.1 | ||||
XM_011529588.2 | Intron | XP_011527890.1 | ||||
XM_011529590.2 | Intron | XP_011527892.1 | ||||
XM_017028360.1 | Intron | XP_016883849.1 | ||||
XM_017028361.1 | Intron | XP_016883850.1 |
ZNF295-AS1 - ZNF295 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |