Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MGAM - maltase-glucoamylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004668.2 | Intron | NP_004659.2 | ||||
XM_006716168.2 | Intron | XP_006716231.1 | ||||
XM_011516670.2 | Intron | XP_011514972.1 | ||||
XM_011516671.2 | Intron | XP_011514973.1 | ||||
XM_011516672.2 | Intron | XP_011514974.1 | ||||
XM_011516673.2 | Intron | XP_011514975.1 | ||||
XM_011516674.2 | Intron | XP_011514976.1 | ||||
XM_017012772.1 | Intron | XP_016868261.1 |
Set Membership: |
HapMap |