Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACAAAAGGCATCTCTTTTCTCT[C/T]TCTTATGCACCAAGCAGAGTATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604987 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLEC5A - C-type lectin domain family 5 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301167.1 | 1851 | UTR 3 | NP_001288096.1 | |||
NM_013252.2 | 1851 | UTR 3 | NP_037384.1 | |||
XM_011515995.2 | 1851 | UTR 3 | XP_011514297.1 | |||
XM_017011915.1 | 1851 | UTR 3 | XP_016867404.1 | |||
XM_017011916.1 | 1851 | Intron | XP_016867405.1 | |||
XM_017011917.1 | 1851 | UTR 3 | XP_016867406.1 |
OR9A4 - olfactory receptor family 9 subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |