Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165080 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ERG - ERG, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136154.1 | Intron | NP_001129626.1 | ||||
NM_001136155.1 | Intron | NP_001129627.1 | ||||
NM_001243428.1 | Intron | NP_001230357.1 | ||||
NM_001243429.1 | Intron | NP_001230358.1 | ||||
NM_001243432.2 | Intron | NP_001230361.1 | ||||
NM_001291391.1 | Intron | NP_001278320.1 | ||||
NM_004449.4 | Intron | NP_004440.1 | ||||
NM_182918.3 | Intron | NP_891548.1 | ||||
XM_011529486.2 | Intron | XP_011527788.1 | ||||
XM_011529487.2 | Intron | XP_011527789.1 | ||||
XM_017028287.1 | Intron | XP_016883776.1 | ||||
XM_017028288.1 | Intron | XP_016883777.1 | ||||
XM_017028289.1 | Intron | XP_016883778.1 |
Set Membership: |
HapMap Validated |