Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCATAATAAATTAACTTAAAAA[A/C]AAAGAGAGCATATTTTGACTAAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604347 | ||||||||||||||||||||
Literature Links: |
MIR186 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR186 - microRNA 186 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZRANB2 - zinc finger RANBP2-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005455.4 | Intron | NP_005446.2 | ||||
NM_203350.2 | Intron | NP_976225.1 |
ZRANB2-AS1 - ZRANB2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |