Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300103 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHROOM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SHROOM2 - shroom family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320663.1 | Intron | NP_001307592.1 | ||||
NM_001320664.1 | Intron | NP_001307593.1 | ||||
NM_001649.3 | Intron | NP_001640.1 | ||||
XM_005274500.4 | Intron | XP_005274557.2 | ||||
XM_011545506.1 | Intron | XP_011543808.1 | ||||
XM_017029492.1 | Intron | XP_016884981.1 | ||||
XM_017029493.1 | Intron | XP_016884982.1 | ||||
XM_017029494.1 | Intron | XP_016884983.1 |
Set Membership: |
HapMap |