Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125290 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALAS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ALAS1 - 5'-aminolevulinate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000688.5 | 46 | UTR 5 | NP_000679.1 | |||
NM_001304443.1 | 46 | UTR 5 | NP_001291372.1 | |||
NM_001304444.1 | 46 | UTR 5 | NP_001291373.1 | |||
NM_199166.2 | 46 | UTR 5 | NP_954635.1 | |||
XM_011533477.1 | 46 | UTR 5 | XP_011531779.1 | |||
XM_011533478.2 | 46 | UTR 5 | XP_011531780.1 | |||
XM_017005873.1 | 46 | UTR 5 | XP_016861362.1 |
Set Membership: |
HapMap |