Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176977 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRKCD - protein kinase C delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316327.1 | Intron | NP_001303256.1 | ||||
NM_006254.3 | Intron | NP_006245.2 | ||||
NM_212539.1 | Intron | NP_997704.1 | ||||
XM_006713257.3 | Intron | XP_006713320.1 | ||||
XM_006713259.2 | Intron | XP_006713322.1 | ||||
XM_017006855.1 | Intron | XP_016862344.1 | ||||
XM_017006856.1 | Intron | XP_016862345.1 |
Set Membership: |
Validated |