Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176977 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRKCD - protein kinase C delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316327.1 | 376 | Intron | NP_001303256.1 | |||
NM_006254.3 | 376 | Intron | NP_006245.2 | |||
NM_212539.1 | 376 | Intron | NP_997704.1 | |||
XM_006713257.3 | 376 | Intron | XP_006713320.1 | |||
XM_006713259.2 | 376 | Intron | XP_006713322.1 | |||
XM_017006855.1 | 376 | Intron | XP_016862344.1 | |||
XM_017006856.1 | 376 | UTR 5 | XP_016862345.1 |
Set Membership: |
HapMap |