Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607612 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLSCR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLSCR4 - phospholipid scramblase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128304.1 | Intron | NP_001121776.1 | ||||
NM_001128305.1 | Intron | NP_001121777.1 | ||||
NM_001128306.1 | Intron | NP_001121778.1 | ||||
NM_001177304.1 | Intron | NP_001170775.1 | ||||
NM_020353.2 | Intron | NP_065086.2 | ||||
XM_005247654.2 | Intron | XP_005247711.1 | ||||
XM_005247655.2 | Intron | XP_005247712.1 | ||||
XM_011513031.2 | Intron | XP_011511333.1 | ||||
XM_017006921.1 | Intron | XP_016862410.1 |
Set Membership: |
HapMap |